Ethyl-4-phenylpyrrole-3-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

B24877-03 B24877-06 B24877-14
CAAAAB24877-03EA 130.67 CAD
CAAAAB24877-03 CAAAAB24877-06 CAAAAB24877-14
Ethyl-4-phenylpyrrole-3-carboxylate 97%
Ethyl-4-phenylpyrrole-3-carboxylate

Purity: 97% CAS No.: 64276-62-6 Molecular Formula: C13H13NO2 Formula Weight: 215.25 Melting Point: 126-128° MDL No.: MFCD03931521

Formula: C₁₃H₁₃NO₂
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD03931521
CAS Number: 64276-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR