1-Chloro-3,5-dimethoxybenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

B24834-06 B24834-14 B24834-22
CAAAAB24834-06EA 67.62 CAD
CAAAAB24834-06 CAAAAB24834-14 CAAAAB24834-22
1-Chloro-3,5-dimethoxybenzene 98%
1-Chloro-3,5-dimethoxybenzene

Purity: 98% CAS No.: 7051-16-3 Molecular Formula: C8H9ClO2 Formula Weight: 172.61 Flash Point: 112°(233°F) Melting Point: 32-36° Boiling Point: 234° Beilstein Reference: 2327304 MDL No.: MFCD00008382

Formula: C₈H₉ClO₂
Boiling Pt: 234 °C
Melting Pt: 32…36 °C
Flash Pt: 112 °C(233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008382
CAS Number: 7051-16-3
EINECS: 230-330-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR