2-Hydroxyethyl macrylate 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 200-300 ppm

B24815-0B B24815-30
CAAAAB24815-0BEA 254.38 CAD
CAAAAB24815-0B CAAAAB24815-30
2-Hydroxyethyl macrylate 97% stabilized
2-Hydroxyethyl macrylate

As a reactive monomer


Purity: 97% CAS No.: 818-61-1 Molecular Formula: C5H8O3 Formula Weight: 116.12 Flash Point: 101°(214°F) Refractive Index: 1.4500 Odor: Pungent Melting Point: -60° Boiling Point: 192° UN No.: 2927 MDL No.: MFCD00002865

Formula: C₅H₈O₃
Boiling Pt: 192 °C
Melting Pt: < –60 °C
Density: 1.106
Flash Pt: 101 °C(214 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002865
CAS Number: 818-61-1
EINECS: 212-454-9
UN: 2927
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR