Ethyl hippurate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl Hippurate, Ethyl 2-benzamidoacetate, Ethyl 2-(phenylformamido)acetate

B24765-06 B24765-14
CAAAAB24765-06EA 113.76 CAD
CAAAAB24765-06 CAAAAB24765-14 CAAAAB24765-22
Ethyl hippurate 98%
Ethyl hippurate

Purity: 98% CAS No.: 1499-53-2 Molecular Formula: C6H5CONHCH2CO2CH2CH3 Formula Weight: 207.23 Melting Point: 58-60° MDL No.: MFCD00026890

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00026890
CAS Number: 1499-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR