(Benzyl)triphenylphosphonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

B24567-09 B24567-18 B24567-30
CAAAAB24567-09EA 42.82 CAD
CAAAAB24567-09 CAAAAB24567-18 CAAAAB24567-30
(Benzyl)triphenylphosphonium bromide 98%
(Benzyl)triphenylphosphonium bromide

Purity: 98% CAS No.: 1449-46-3 Molecular Formula: C6H5CH2P(C6H5)3Br Formula Weight: 433.34 Melting Point: 293-298° Beilstein Reference: 3599867 UN No.: 3464 MDL No.: MFCD00031707

Formula: C₂₅H₂₂BrP
Melting Pt: 293…298 °C
Storage Temperature: Ambient
MDL Number: MFCD00031707
CAS Number: 1449-46-3
EINECS: 215-908-4
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR