(±)-Pipecolinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

B23541-06 B23541-14 B23541-22
CAAAAB23541-06EA 62.57 CAD
CAAAAB23541-06 CAAAAB23541-14 CAAAAB23541-22
(±)-Pipecolinic acid 99%
(±)-Pipecolinic acid

Purity: 99% CAS No.: 535-75-1 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Melting Point: ca 280° dec. Beilstein Reference: 81095 Merck Reference: 14,7458 MDL No.: MFCD00064347

Formula: C₆H₁₁NO₂
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064347
CAS Number: 535-75-1
EINECS: 208-616-3
Merck Index: 14,7458

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR