Thymol blue sodium salt

Supplier: Thermo Scientific Chemicals

B23007-09 B23007-18 B23007-30
CAAAAB23007-09EA 63.99 CAD
CAAAAB23007-09 CAAAAB23007-18 CAAAAB23007-30
Thymol blue sodium salt
Thymol blue sodium salt

CAS No.: 62625-21-2 Molecular Formula: C27H29NaO5S Formula Weight: 488.58 Melting Point: ca 284° dec. Beilstein Reference: 3861515 MDL No.: MFCD00151093

Formula: C₂₇H₂₉NaO₅S
Melting Pt: ∼ 284 °C (dec.)
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151093
CAS Number: 62625-21-2
EINECS: 263-650-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR