Phenol red

Supplier: Thermo Scientific Chemicals
Warning

B21710-09 B21710-18 B21710-30
CAAAAB21710-09EA 62.73 CAD
CAAAAB21710-09 CAAAAB21710-18 CAAAAB21710-30
Phenol red
Phenol red

CAS No.: 143-74-8 Molecular Formula: C19H14O5S Formula Weight: 354.38 Odor: Odorless Boiling Point: 300° Beilstein Reference: 326470 Merck Reference: 14,7247 MDL No.: MFCD00003552

Formula: C₁₉H₁₄O₅S
Boiling Pt: >300 °C
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Merck Index: 14,7247

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR