Dihydrocoumarin 99%

Supplier: Thermo Scientific Chemicals
Warning

B21229-0E B21229-22 B21229-36
CAAAAB21229-0EEA 644.64 CAD
CAAAAB21229-0E CAAAB21229-22 CAAAB21229-36
Dihydrocoumarin 99%
Dihydrocoumarin

Purity: 99% CAS No.: 119-84-6 Molecular Formula: C9H8O2 Formula Weight: 148.16 Flash Point: 130°(266°F) Refractive Index: 1.5560 Melting Point: 24-25° Boiling Point: 272° Beilstein Reference: 4584 MDL No.: MFCD00006881

Formula: C₉H₈O₂
Boiling Pt: 272 °C
Melting Pt: 24…25 °C
Density: 1.169
Flash Pt: 130 °C(266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006881
CAS Number: 119-84-6
EINECS: 204-354-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR