3,5-Dichlorosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

B20941-22 B20941-06 B20941-14
CAAAAB20941-22EA 511.92 CAD
CAAAAB20941-22 CAAAB20941-06 CAAAB20941-14
3,5-Dichlorosalicylaldehyde 98%
3,5-Dichlorosalicylaldehyde

Purity: 98% CAS No.: 90-60-8 Molecular Formula: C7H4Cl2O2 Formula Weight: 191.01 Melting Point: 94-97° Beilstein Reference: 973391

Formula: C₇H₄Cl₂O₂
Boiling Pt: 723 °C (1013 hPa)
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00003320
CAS Number: 90-60-8
EINECS: 202-005-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR