Aluminium trifluoromethanesulfonate 99%

Supplier: Thermo Scientific Chemicals
Danger

B20785-06 B20785-14
CAAAAB20785-06EA 96.85 CAD
CAAAAB20785-06 CAAAAB20785-14
Aluminium trifluoromethanesulfonate 99%
Aluminium trifluoromethanesulphonate

Purity: 99% CAS No.: 74974-61-1 Molecular Formula: C3AlF9O9S3 Formula Weight: 474.20 Melting Point: ca 300° UN No.: 3261 MDL No.: MFCD00143596

Formula: C₃AlF₉O₉S₃
Melting Pt: ca 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00143596
CAS Number: 74974-61-1
UN: 3261
ADR: "8",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR