2,2,3,4,4,4-Hexafluoro-1-butanol 95%

Supplier: Thermo Scientific Chemicals
Warning

B20587-22 B20587-06 B20587-14
CAAAAB20587-22EA 567.22 CAD
CAAAAB20587-22 CAAAB20587-06 CAAAB20587-14
2,2,3,4,4,4-Hexafluoro-1-butanol 95%
2,2,3,4,4,4-Hexafluoro-1-butanol

Purity: 95% CAS No.: 382-31-0 Molecular Formula: CF3CHFCF2CH2OH Formula Weight: 182.07 Flash Point: 51°(123°F) Refractive Index: 1.3120 Boiling Point: 114-116° Beilstein Reference: 1753872 UN No.: 1993 MDL No.: MFCD00042309

Formula: C₄H₄F₆O
Boiling Pt: 114…116 °C
Density: 1.64
Flash Pt: 51 °C (123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042309
CAS Number: 382-31-0
EINECS: 206-842-7
UN: 1993
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR