Nitronium tetrafluoroborate 96%

Supplier: Thermo Scientific Chemicals
Danger

B20167-06 B20167-14 B20167-22
CAAAAB20167-06EA 102.7 CAD
CAAAAB20167-06 CAAAAB20167-14 CAAAAB20167-22
Nitronium tetrafluoroborate 96%
Nitronium tetrafluoroborate

Purity: 96% CAS No.: 13826-86-3 Molecular Formula: BF4NO2 Formula Weight: 132.81 Storage Temperature: 2°C to 8°C Melting Point: ca 240° dec. UN No.: 3260 MDL No.: MFCD00011432

Formula: BF₄NO₂
Melting Pt: ∼ 240 °C (dec.)
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011432
CAS Number: 13826-86-3
EINECS: 237-533-5
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR