trans-Aconitic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20087-14 B20087-22
CAAAAB20087-14EA 93.54 CAD
CAAAAB20087-14 CAAAAB20087-22
trans-Aconitic acid 98%
trans-Aconitic acid

Purity: 98% CAS No.: 4023-65-8 Molecular Formula: C6H6O6 Formula Weight: 174.11 Odor: Odorless Melting Point: ca 190° dec. MDL No.: MFCD00002721

Formula: C₆H₆O₆
Melting Pt: ca 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002721
CAS Number: 4023-65-8
EINECS: 223-688-6
Merck Index: 14,117

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR