(±)-1,2-Propanediol 98+%

Supplier: Thermo Scientific Chemicals

A18406-0E A18406-36
CAAAAA18406-0EEA 140.46 CAD
CAAAAA18406-0E CAAAAA18406-36
(±)-1,2-Propanediol 98+%
Propylene glycol

Purity: 98+% CAS No.: 57-55-6 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: 107°(225°F) Refractive Index: 1.4320 Odor: Nearly odorless pH: neutral Melting Point: -60° Boiling Point: 186-188° Beilstein Reference: 1340498 Merck Reference: 14,7855 MDL No.: MFCD00064272

Formula: CH₃CH(OH)CH₂OH
Boiling Pt: 186…188 °C
Melting Pt: –60 °C
Density: 1.036
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 14,7855
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR