Isonicotinamide 99%

Supplier: Thermo Scientific Chemicals
Warning

A18190-22 A18190-36 A18190-18 A18190-30
CAAAAA18190-22EA 99.07 CAD
CAAAAA18190-22 CAAAAA18190-36 CAAAA18190-18 CAAAA18190-30
Isonicotinamide 99%
Isonicotinamide

Purity: 99% CAS No.: 1453-82-3 Molecular Formula: C6H6N2O Formula Weight: 122.13 Melting Point: 155-158° Beilstein Reference: 2173

Formula: C₆H₆N₂O
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00006432
CAS Number: 1453-82-3
EINECS: 215-926-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR