Gibberellic acid 90+%

Supplier: Thermo Scientific Chemicals
Warning

A17843-03 A17843-06 A17843-14
CAAAAA17843-03EA 82.32 CAD
CAAAAA17843-03 CAAAAA17843-06 CAAAAA17843-14
Gibberellic acid 90+%
Gibberellic acid

Plant growth hormone


Purity: 90+% CAS No.: 77-06-5 Molecular Formula: C19H22O6 Formula Weight: 346.37 Melting Point: 222° dec. Beilstein Reference: 54346 Merck Reference: 14,4419 Optical Rotation: +78° (c=2 in methanol) MDL No.: MFCD00079329

Formula: C₁₉H₂₂O₆
Melting Pt: 222 °C (dec.)
Density: 1.386 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079329
CAS Number: 77-06-5
EINECS: 201-001-0
Merck Index: 14,4419

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR