Copper(II) bromide 99%

Supplier: Thermo Scientific Chemicals
Danger

A16335-14 A16335-22 A16335-36
CAAAAA16335-14EA 55.93 CAD
CAAAAA16335-14 CAAAAA16335-22 CAAAAA16335-36
Copper(II) bromide 99%
Copper (II) bromide

Purity: 99% CAS No.: 7789-45-9 Molecular Formula: CuBr2 Formula Weight: 223.36 Odor: Odorless Melting Point: 498° Boiling Point: 900° Merck Reference: 14,2629 UN No.: 3260 MDL No.: MFCD00010970

Formula: Br₂Cu
Boiling Pt: 900 °C
Melting Pt: 498 °C
Density: 4.710
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8,III
Merck Index: 14,2629

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR