Tin(II) chloride, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Stannous chloride, Tin dichloride

A16202-0E A16202-22 A16202-36
CAAAAA16202-0EEA 973.28 CAD
CAAAAA16202-0E CAAAAA16202-22 CAAAAA16202-36
Tin(II) chloride, anhydrous 98%
Tin (II) chloride

Purity: 98% CAS No.: 7772-99-8 Molecular Formula: SnCl2 Formula Weight: 189.62 Melting Point: 247° Boiling Point: 652° Merck Reference: 14,8783 UN No.: 3260 MDL No.: MFCD00011241

Formula: SnCl₂
MW: 189.62 g/mol
Boiling Pt: 623
Melting Pt: 246
Density: 3.95
Storage Temperature: Ambient
MDL Number: MFCD00011241
CAS Number: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 14,8783

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR