Sodium nitroprusside dihydrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium pentacyanonitrosylferrate(III) dihydrate, Sodium nitrosopentacyanoferrate (III) dihydrate, Sodium nitroferricyanide(III) dihydrate, Disodium pentacyanonitrosylferrate (III) dihydrate, Sodium nitroferricyanide dihydrate

A15656-14 A15656-22 A15656-36
CAAAAA15656-14EA 56.88 CAD
CAAAAA15656-14 CAAAAA15656-22 CAAAAA15656-36
Sodium nitroprusside dihydrate 98+%
Sodium nitroprusside dihydrate

Purity: 98+% CAS No.: 13755-38-9 Molecular Formula: C5FeN6Na2O.2H2O Formula Weight: 297.95 (261.92anhy) Odor: Odorless Merck Reference: 14,8649 MDL No.: MFCD00149192

Formula: Na₂[Fe(CN)₅NO]·2H₂O
Density: 1.72
Storage Temperature: Ambient
MDL Number: MFCD00149192
CAS Number: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,III
Merck Index: 14,8649

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR