p-Terphenyl 99+%

Supplier: Thermo Scientific Chemicals
Warning

A14833-30 A14833-14 A14833-22
CAAAAA14833-30EA 323.9 CAD
CAAAAA14833-30 CAAAA14833-14 CAAAA14833-22
p-Terphenyl 99+%
p-Terphenyl

Purity: 99+% CAS No.: 92-94-4 Molecular Formula: C18H14 Formula Weight: 230.31 Flash Point: 207°(404°F) Melting Point: 212-215° Boiling Point: 405° Beilstein Reference: 1908447 MDL No.: MFCD00003061

Formula: C₁₈H₁₄
Boiling Pt: 405 °C
Melting Pt: 212…215 °C
Density: 1.23
Flash Pt: 207 °C(404 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003061
CAS Number: 92-94-4
EINECS: 202-205-2
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR