4,4-Dimethyl-2-oxazoline 98%

Supplier: Thermo Scientific Chemicals
Danger

A12670-22 A12670-06 A12670-14
CAAAAA12670-22EA 1072.82 CAD
CAAAAA12670-22 CAAAA12670-06 CAAAA12670-14
4,4-Dimethyl-2-oxazoline 98%
4,4-Dimethyl-2-oxazoline

Purity: 98% CAS No.: 30093-99-3 Molecular Formula: C5H9NO Formula Weight: 99.13 Flash Point: 1°(33°F) Refractive Index: 1.4190 Boiling Point: 99-100° Beilstein Reference: 1209606 UN No.: 1993 MDL No.: MFCD00014507

Formula: C₅H₉NO
Boiling Pt: 99…100 °C
Density: 0.940
Flash Pt: 1 °C(33 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014507
CAS Number: 30093-99-3
EINECS: 250-045-7
UN: 1993
ADR: "3",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR