Ethyl-4-pentenoate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

A12422-03 A12422-22
CAAAAA12422-03EA 50.88 CAD
CAAAAA12422-03 CAAAA12422-22
Ethyl-4-pentenoate ≥98%
Ethyl-4-pentenoate

Purity: 98+% CAS No.: 1968-40-7 Molecular Formula: CH2=CHCH2CH2CO2CH2CH3 Formula Weight: 128.17 Flash Point: 37°(98°F) Refractive Index: 1.4170 Boiling Point: 146° Beilstein Reference: 1751643 UN No.: 3272 MDL No.: MFCD00036562

Formula: C₇H₁₂O₂
Boiling Pt: 146 °C
Density: 0.900
Flash Pt: 37 °C (98 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036562
CAS Number: 1968-40-7
EINECS: 217-818-0
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR