Ethyl tiglate 98%

Supplier: Thermo Scientific Chemicals
Warning

A12029-14 A12029-22
CAAAAA12029-14EA 78.68 CAD
CAAAAA12029-14 CAAAAA12029-22
Ethyl tiglate 98%
Ethyl tiglate

Purity: 98% CAS No.: 5837-78-5 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 46°(114°F) Refractive Index: 1.4340 Boiling Point: 156° Beilstein Reference: 1720895 Merck Reference: 14,9433 UN No.: 3272 MDL No.: MFCD00015183

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 156 °C
Density: 0.930
Flash Pt: 46 °C (114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015183
CAS Number: 5837-78-5
EINECS: 227-425-6
UN: 3272
ADR: 3 III
Merck Index: 14,9433

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR