(±)-2-Methylbutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A11546-AC A11546-0F A11546-AP
CAAAAA11546-ACEA 40.45 CAD
CAAAAA11546-AC CAAAA11546-0F CAAAA11546-AP
(±)-2-Methylbutyric acid 98%
(±)-2-Methylbutyric acid

MDL: MFCD00002669 Beilstein Registry No.: 1720486


Purity: 98% CAS No.: 116-53-0 Molecular Formula: C5H10O2 Formula Weight: 102.14 Flash Point: 73°(163°F) Physical Form: Liquid Density: 0.937 Refractive Index: 1.4055 Boiling Point: 175-176° Beilstein Reference: 1720486 UN No.: 3265 MDL No.: MFCD00002669

Formula: C₅H₁₀O₂
Boiling Pt: 175…176 °C
Melting Pt: –70 °C
Density: 0.937
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002669
CAS Number: 116-53-0
EINECS: 204-145-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR