3-Iodobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11515-30 A11515-09 A11515-18
CAAAAA11515-30EA 649.38 CAD
CAAAAA11515-30 CAAAA11515-09 CAAAA11515-18
3-Iodobenzoic acid 98+%
3-Iodobenzoic acid

MDL: MFCD00002496 Beilstein Registry No.: 971088


Purity: 98+% CAS No.: 618-51-9 Molecular Formula: C7H5IO2 Formula Weight: 248.02 Melting Point: 186-188° Beilstein Reference: 971088

Formula: C₇H₅IO₂
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 186…188 °C
Density: 2,171 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002496
CAS Number: 618-51-9
EINECS: 210-555-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR