2-Amino-4,6-dichloropyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

A11330-22 A11330-06 A11330-14
CAAAAA11330-22EA 862.68 CAD
CAAAAA11330-22 CAAAA11330-06 CAAAA11330-14
2-Amino-4,6-dichloropyrimidine 98%
2-Amino-4,6-dichloropyrimidine

MDL: MFCD00006090 Beilstein Registry No.: 118454


Purity: 98% CAS No.: 56-05-3 Molecular Formula: C4H3Cl2N3 Formula Weight: 164.00 Melting Point: 218-222° Beilstein Reference: 118454

Formula: C₄H₃Cl₂N₃
Melting Pt: 218…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00006090
CAS Number: 56-05-3
EINECS: 200-253-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR