L(-)-Tyrosine 99%

Supplier: Thermo Scientific Chemicals
Warning

A11141-22 A11141-0B A11141-18 A11141-30
CAAAAA11141-22EA 111.39 CAD
CAAAAA11141-22 CAAAA11141-0B CAAAA11141-18 CAAAA11141-30
L(-)-Tyrosine 99%
L(-)-Tyrosine

MDL: MFCD00002606 Beilstein Registry No.: 392441 Optical Rotation: -11°C (c=4 in 1N HCl)


Purity: 99% CAS No.: 60-18-4 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Physical Form: Cryst powder Density: 1.46 Appearance: White Melting Point: 300° Beilstein Reference: 392441 Merck Reference: 14,9839 Optical Rotation: -11° (c=4 in 1N HCl) MDL No.: MFCD00002606

Formula: C₉H₁₁NO₃
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: >300 °C
Density: 1.46
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
EINECS: 200-460-4
Merck Index: 14,9839

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR