Antimony(III) oxide 99%

Supplier: Thermo Scientific Chemicals
Warning

A11123-0E A11123-22 A11123-36
CAAAAA11123-0EEA 431.34 CAD
CAAAAA11123-0E CAAAAA11123-22 CAAAAA11123-36
Antimony(III) oxide 99%
Antimony (III) oxide

Purity: 99% CAS No.: 1309-64-4 Molecular Formula: Sb2O3 Formula Weight: 291.50 Odor: Odorless Melting Point: 656° Boiling Point: 1550° subl. Merck Reference: 14,711 MDL No.: MFCD00011214

Formula: O₃Sb₂
Boiling Pt: 1550 °C (subl.)
Melting Pt: 656 °C
Density: 5.67
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS Number: 1309-64-4
EINECS: 215-175-0
ADR: 6.1,III
Merck Index: 14,711

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR