D-(+)-Camphor 98%

Supplier: Thermo Scientific Chemicals
Warning

A10708-14 A10708-22 A10708-36
CAAAAA10708-14EA 42.19 CAD
CAAAAA10708-14 CAAAAA10708-22 CAAAAA10708-36
D-(+)-Camphor 98%
D(+)-Camphor

Purity: 98% CAS No.: 464-49-3 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 65°(149°F) Odor: Camphor-like Boiling Point: 204° Beilstein Reference: 2042745 Merck Reference: 14,1732 Optical Rotation: +43° (c=10 in ethanol) UN No.: 2717 MDL No.: MFCD00064149

Formula: C₁₀H₁₆O
Boiling Pt: 204 °C
Melting Pt: 175…180 °C
Density: 0.990
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064149
CAS Number: 464-49-3
EINECS: 207-355-2
UN: 2717
ADR: 4.1,III
Merck Index: 14,1732

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR