Sodium perborate tetrahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

A10189-0E A10189-22 A10189-36
CAAAAA10189-0EEA 127.35 CAD
CAAAAA10189-0E CAAAAA10189-22 CAAAAA10189-36
Sodium perborate tetrahydrate 97%
Sodium perborate tetrahydrate

Purity: 97% CAS No.: 10486-00-7 Molecular Formula: NaBO3.4H2O Formula Weight: 153.86 (81.84anhy) Odor: Odorless pH: ca 10 Melting Point: 60° dec. Merck Reference: 14,8652 UN No.: 1479 MDL No.: MFCD00149231

Formula: NaH₂BO₄·4H₂O
Melting Pt: 60 °C (dec.)
Density: 1.73
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9
UN: 3377
ADR: 5.1,III
Merck Index: 14,8652

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR