Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III) 99%

Supplier: Thermo Scientific Chemicals
Warning

44842-03
CAAAA44842-03EA 128.3 CAD
CAAAA44842-03 CAAAA44842-06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III) 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III)

Granular


Purity: 99% CAS No.: 15492-51-0 Molecular Formula: C33H57O6Tb Formula Weight: 708.74 Physical Form: Granular Melting Point: 155-156° MDL No.: MFCD00064752

Formula: C₃₃H₅₇O₆Tb
Melting Pt: 155…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00064752
CAS Number: 15492-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR