Silicon(IV) 2-ethylhexanoate ≥75% in ethanol

Supplier: Thermo Scientific Chemicals
Danger

44635-30
CAAAA44635-30EA 862.68 CAD
CAAAA44635-30
Silicon(IV) 2-ethylhexanoate ≥75% in ethanol
Silicon(IV) 2-ethylhexanoate

Liquid


CAS No.: 67939-81-5 Molecular Formula: C32H60O8Si Formula Weight: 600.91 Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Yellow UN No.: 1170 MDL No.: MFCD00269842

Formula: C₃₂H₆₀O₈Si
Storage Temperature: Ambient
MDL Number: MFCD00269842
CAS Number: 67939-81-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR