Silver perrhenate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

44480-03 44480-06
CAAAA44480-03EA 147.26 CAD
CAAAA44480-03 CAAAA44480-06 CAAAA44480-14
Silver perrhenate ≥99.99% (metals basis)
Silver perrhenate

Purity: 99.99% CAS No.: 20654-56-2 Molecular Formula: AgO4Re Formula Weight: 358.10 Physical Form: Powder Density: 7.05 Appearance: White Melting Point: 430° UN No.: 1479 MDL No.: MFCD00014144

Formula: AgO₄Re
Melting Pt: 430 °C
Density: 7.05
Storage Temperature: Ambient
MDL Number: MFCD00014144
CAS Number: 20654-56-2
EINECS: 232-042-2
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR