Zinc oxide ≥99.9% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

41126-KS
CAAAA41126-KSEA 2253.08 CAD
CAAAA41126-KS
Zinc oxide ≥99.9% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Zinc oxide

Purity: 99.9% Molecular Formula: ZnO Formula Weight: 81.37 MDL No.: MFCD00011300

Formula: ZnO
MW: 81.39 g/mol
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR