Titanium nitride ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals

41111-KS
CAAAA41111-KSEA 1714.3 CAD
CAAAA41111-KS
Titanium nitride ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Titanium nitride

Purity: 99.5% CAS No.: 25583-20-4 Molecular Formula: TiN Formula Weight: 61.91 Physical Form: Hot pressed material Density: 5.22 Appearance: Gray Melting Point: 2950° MDL No.: MFCD00049596

Formula: NTi
Melting Pt: 2950 °C
Density: 5.22
Storage Temperature: Ambient
MDL Number: MFCD00049596
CAS Number: 25583-20-4
EINECS: 247-117-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR