N-Cbz-L-serine 99%

Supplier: Thermo Scientific Chemicals

A19050-22 A19050-06 A19050-14
CAAAAA19050-22EA 470.84 CAD
CAAAAA19050-22 CAAAA19050-06 CAAAA19050-14
N-Cbz-L-serine 99%
N-Cbz-L-serine

Purity: 99% CAS No.: 1145-80-8 Molecular Formula: C11H13NO5 Formula Weight: 239.23 Melting Point: 117-120° Beilstein Reference: 2058314 Optical Rotation: +6° (c=7 in acetic acid) MDL No.: MFCD00002662

Formula: C11H13NO5
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002662
CAS Number: 1145-80-8
EINECS: 214-546-4

Order Now

Specification Test Results

Optical rotation +6° (c=7 in acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR