2-Acrylamido-2-methylpropanesulfonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A18924-0B A18924-30
CAAAA18924-0BEA 238.58 CAD
CAAAA18924-0B CAAAA18924-30
2-Acrylamido-2-methylpropanesulfonic acid 98%
2-Acrylamido-2-methylpropanesulphonic acid

Purity: 98% CAS No.: 15214-89-8 Molecular Formula: C7H13NO4S Formula Weight: 207.25 Storage Temperature: 2°C to 8°C Melting Point: ca 195° dec. Beilstein Reference: 1946464 UN No.: 2585 MDL No.: MFCD00007522

Formula: C₇H₁₃NO₄S
Melting Pt: ca 195 °C (dec.)
Density: 1.10
Storage Temperature: Refrigerator
MDL Number: MFCD00007522
CAS Number: 15214-89-8
EINECS: 239-268-0
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR