Acetylenedicarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

A18736-06 A18736-14 A18736-22
CAAAA18736-06EA 54.35 CAD
CAAAA18736-06 CAAAA18736-14 CAAAA18736-22
Acetylenedicarboxylic acid 97%
Acetylenedicarboxylic acid

Fieser: 1,12


Purity: 97% CAS No.: 142-45-0 Molecular Formula: HO2CC≡CCO2H Formula Weight: 114.06 Storage Temperature: 2°C to 8°C Flash Point: 131°(267°F) Melting Point: ca 182° dec. Beilstein Reference: 878357 UN No.: 2928 MDL No.: MFCD00004362

Formula: C₄H₂O₄
Melting Pt: ca 182 °C (dec.)
Flash Pt: 131 °C(267 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004362
CAS Number: 142-45-0
EINECS: 205-536-0
UN: 2811
ADR: "6.1",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR