1-Bromo-2,5-difluoro-4-nitrobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

A18670-06
CAAAA18670-03EA 109.73 CAD
CAAAA18670-03 CAAAA18670-06
1-Bromo-2,5-difluoro-4-nitrobenzene 97%
1-Bromo-2,5-difluoro-4-nitrobenzene

Purity: 97% CAS No.: 167415-27-2 Molecular Formula: C6H2BrF2NO2 Formula Weight: 237.99 Melting Point: 55-57° Beilstein Reference: 7411347

Formula: C₆H₂BrF₂NO₂
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00042184
CAS Number: 167415-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR