1,10-Phenanthroline-2,9-dicarboxylic acid hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

A18548-01 A18548-03
CAAAA18548-01EA 274.92 CAD
CAAAA18548-01 CAAAA18548-03
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate 98%
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate

Purity: 98% CAS No.: 57709-61-2 Molecular Formula: C14H8N2O4.xH2O Formula Weight: 268.23(anhy) MDL No.: MFCD00209670

Formula: C₁₄H₈N₂O₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00209670
CAS Number: 57709-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR