7-Hydroxyflavone 98%

Supplier: Thermo Scientific Chemicals

A18538-03 A18538-06
CAAAA18538-03EA 85.16 CAD
CAAAA18538-03 CAAAA18538-06
7-Hydroxyflavone 98%
7-Hydroxyflavone

Purity: 98% CAS No.: 6665-86-7 Molecular Formula: C15H10O3 Formula Weight: 238.25 Melting Point: 245-247° Beilstein Reference: 194089

Formula: C₁₅H₁₀O₃
Boiling Pt: 395 °C (1013 hPa)
Melting Pt: 245…247 °C
Storage Temperature: Ambient
MDL Number: MFCD00006835
CAS Number: 6665-86-7
EINECS: 229-705-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR