Glutaconaldehydedianil hydrochloride mixture of isomers 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(5-Anilino-2,4-pentadienylidene)aniline hydrochloride, N-[5-(Phenylamino)-2,4-pentadienylidene]aniline monohydrochloride, N-((2E,4E)-5-(Phenylamino)penta-2,4-dien-1-ylidene)aniline hydrochloride

A18390-22 A18390-06 A18390-14
CAAAAA18390-22EA 1540.5 CAD
CAAAAA18390-22 CAAAA18390-06 CAAAA18390-14
Glutaconaldehydedianil hydrochloride mixture of isomers 98%
Glutaconaldehydedianil hydrochloride

Purity: 98% CAS No.: 1497-49-0 Molecular Formula: C6H5NHCH=CHCH=CHCH=NC6H5.HCl Formula Weight: 284.79 Melting Point: ca 124-132° Beilstein Reference: 3916924 MDL No.: MFCD00012630

Formula: C₁₇H₁₆N₂
MW: 284.79 g/mol
Melting Pt: ∼ 124…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00012630
CAS Number: 1497-49-0
EINECS: 216-094-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR