2-Methyl-2-butanol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Amyl alcohol, tert-Pentyl alcohol, tert-Pentanol

A18304-0D A18304-0F A18304-AP
CAAAA18304-0DEA 850.04 CAD
CAAAA18304-0D CAAAA18304-0F CAAAA18304-AP
2-Methyl-2-butanol 98%
2-Methyl-2-butanol

Purity: 98% CAS No.: 75-85-4 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: 20°(68°F) Physical Form: Clear liquid Density: 0.805 Refractive Index: 1.4050 Appearance: Colorless Odor: Unpleasant Melting Point: ca -12° Boiling Point: 101-103° Beilstein Reference: 1361351 Merck Reference: 14,7140 UN No.: 1105 MDL No.: MFCD00004478

Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 101…103 °C
Melting Pt: ∼ 12 °C
Density: 0.805
Flash Pt: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004478
CAS Number: 75-85-4
EINECS: 200-908-9
UN: 1105
ADR: 3 III
Merck Index: 14,7140

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR