2-Amino-6-chloropurine 99%

Supplier: Thermo Scientific Chemicals
Warning

A18195-03 A18195-06 A18195-14
CAAAA18195-03EA 80.74 CAD
CAAAA18195-03 CAAAA18195-06 CAAAA18195-14
2-Amino-6-chloropurine 99%
2-Amino-6-chloropurine

Purity: 99% CAS No.: 10310-21-1 Molecular Formula: C5H4ClN5 Formula Weight: 169.57 Melting Point: 300° Beilstein Reference: 9626

Formula: C₅H₄ClN₅
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00075252
CAS Number: 10310-21-1
EINECS: 233-686-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR