Stearic acid 90+%

Supplier: Thermo Scientific Chemicals

A17673-0B A17673-0I
CAAAA17673-0BEA 55.77 CAD
CAAAA17673-0B CAAAA17673-0I
Stearic acid 90+%
Stearic acid

Purity: 90+% CAS No.: 57-11-4 Molecular Formula: CH3(CH2)16CO2H Formula Weight: 284.48 Flash Point: 196°(384°F) Odor: Odorless Boiling Point: 232°/15mm Beilstein Reference: 608585 Merck Reference: 14,8804 MDL No.: MFCD00002752

Formula: C₁₈H₃₆O₂
Boiling Pt: 232 °C/15 mm
Melting Pt: 67…70 °C
Density: 0.941
Flash Pt: 196 °C(384 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 14,8804
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR