Magnesium trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger

A17591-06 A17591-14 A17591-22
CAAAA17591-06EA 52.46 CAD
CAAAA17591-06 CAAAA17591-14 CAAAA17591-22
Magnesium trifluoromethanesulfonate 98%
Magnesium trifluoromethanesulphonate

Purity: 98% CAS No.: 60871-83-2 Molecular Formula: C2F6MgO6S2 Formula Weight: 322.43 Melting Point: 310° Beilstein Reference: 705578 UN No.: 1759 MDL No.: MFCD00042601

Formula: C₂F₆MgO₆S₂
Melting Pt: >310 °C
Storage Temperature: Ambient
MDL Number: MFCD00042601
CAS Number: 60871-83-2
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR