4-(Trifluoromethoxy)aniline 98%

Supplier: Thermo Scientific Chemicals
Danger

A17208-06 A17208-14
CAAAA17208-06EA 51.98 CAD
CAAAA17208-06 CAAAA17208-14
4-(Trifluoromethoxy)aniline 98%
4-(Trifluoromethoxy)aniline

Purity: 98% CAS No.: 461-82-5 Molecular Formula: C7H6F3NO Formula Weight: 177.13 Flash Point: 81°(178°F) Refractive Index: 1.4630 Odor: Characteristic Boiling Point: 73-75°/10mm Beilstein Reference: 2090209 UN No.: 2810 MDL No.: MFCD00041314

Formula: C₇H₆F₃NO
MW: 177.13
Boiling Pt: 73…75 °C (10 mmHg)
Density: 1.31
Flash Pt: 81 °C (178 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041314
CAS Number: 461-82-5
EINECS: 207-317-5
UN: 2810
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR