L-Lysine ethyl ester dihydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

A16824-14 A16824-22
CAAAA16824-14EA 139.99 CAD
CAAAA16824-14 CAAAA16824-22
L-Lysine ethyl ester dihydrochloride 99%
L-Lysine ethyl ester dihydrochloride

Purity: 99% CAS No.: 3844-53-9 Molecular Formula: C8H18N2O2.2HCl Formula Weight: 247.17 Melting Point: ca 145° dec. Beilstein Reference: 3697099 MDL No.: MFCD00039068

Formula: C₈H₂₀Cl₂N₂O₂
Melting Pt: ca 145 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00039068
CAS Number: 3844-53-9
EINECS: 223-340-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR