1-Iodoperfluorbutane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane

A16671-22 A16671-06 A16671-14
CAAAAA16671-22EA 267.02 CAD
CAAAAA16671-22 CAAAA16671-06 CAAAA16671-14
1-Iodoperfluorbutane 97%
1-Iodoperfluorbutane

Purity: 97% CAS No.: 423-39-2 Molecular Formula: CF3(CF2)3I Formula Weight: 345.93 Refractive Index: 1.3290 Odor: Pungent Boiling Point: 67° Beilstein Reference: 1777546 UN No.: 3082 MDL No.: MFCD00001062

Formula: C₄F₉I
MW: 345.93 g/mol
Boiling Pt: 65
Melting Pt: -2
Density: 2.010
Storage Temperature: Ambient
MDL Number: MFCD00001062
CAS Number: 423-39-2
EINECS: 207-025-8
UN: 3082

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR